Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLR10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319612.1 | NP_001306541.1 | 1 | 2344 | 81 | |
XM_005554670.2 | XP_005554727.1 | 2 | 2582 | 81 | ||
GenBank mRNA | AB445685.1 | - | 1 | 2344 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |