Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005585934.2 | XP_005585991.1 | 3 - 4 | 3052 | 78 | |
XM_005585935.2 | XP_005585992.1 | 3 - 4 | 2760 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |