Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005563554.1 | XP_005563611.1 | 1 - 2 | 343 | 81 | |
XM_005563555.1 | XP_005563612.1 | 1 - 2 | 343 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |