Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283603.1 | NP_001270532.1 | 5 - 6 | 684 | 78 | |
XM_005562600.2 | XP_005562657.1 | 4 - 5 | 1537 | 78 | ||
XM_005562601.2 | XP_005562658.1 | 5 - 6 | 2725 | 78 | ||
XM_005562602.2 | XP_005562659.1 | 6 - 7 | 2791 | 78 | ||
XM_015454393.1 | XP_015309879.1 | 4 - 5 | 1459 | 78 | ||
GenBank mRNA | AB220441.1 | - | 5 - 6 | 684 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |