Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283542.1 | NP_001270471.1 | 8 - 9 | 987 | 82 | |
GenBank mRNA | AB169707.1 | - | 8 - 9 | 987 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |