Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPYL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283656.1 | NP_001270585.1 | 1 - 2 | 944 | 85 | |
GenBank mRNA | AB056806.1 | - | 1 - 2 | 85 | ||
AB169870.1 | - | 1 - 2 | 944 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |