Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283255.1 | NP_001270184.1 | 4 - 5 | 1153 | 96 | |
XM_015450530.1 | XP_015306016.1 | 5 - 6 | 1290 | 96 | ||
XM_015450531.1 | XP_015306017.1 | 5 - 6 | 1269 | 96 | ||
XM_015450532.1 | XP_015306018.1 | 5 - 6 | 1272 | 96 | ||
XM_015450533.1 | XP_015306019.1 | 5 - 6 | 1312 | 96 | ||
XM_015450534.1 | XP_015306020.1 | 5 - 6 | 1309 | 96 | ||
XM_015450535.1 | XP_015306021.1 | 4 - 5 | 1162 | 96 | ||
XM_015450536.1 | XP_015306022.1 | 4 - 5 | 1202 | 96 | ||
XM_015450537.1 | XP_015306023.1 | 4 - 5 | 1340 | 96 | ||
XM_015450538.1 | XP_015306024.1 | 3 - 4 | 1520 | 96 | ||
GenBank mRNA | AB168962.1 | - | 4 - 5 | 1153 | 96 |