Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284635.1 | NP_001271564.1 | 3 - 4 | 312 | 89 | |
XM_005589356.2 | XP_005589413.1 | 3 - 4 | 650 | 89 | ||
XM_015440936.1 | XP_015296422.1 | 3 - 4 | 653 | 89 | ||
XM_015440937.1 | XP_015296423.1 | 3 - 4 | 380 | 89 | ||
GenBank mRNA | AB169584.1 | - | 3 - 4 | 316 | 89 | |
AB170222.1 | - | 3 - 4 | 308 | 89 | ||
AB174290.1 | - | 3 - 4 | 312 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |