Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283999.1 | NP_001270928.1 | 2 - 3 | 423 | 95 | |
XM_005559538.2 | XP_005559595.1 | 2 - 3 | 428 | 95 | ||
XM_005559539.2 | XP_005559596.1 | 2 - 3 | 427 | 95 | ||
XM_015452403.1 | XP_015307889.1 | 2 - 3 | 429 | 95 | ||
XM_015452404.1 | XP_015307890.1 | 2 - 3 | 423 | 95 | ||
XM_015452405.1 | XP_015307891.1 | 2 - 3 | 412 | 95 | ||
XM_015452406.1 | XP_015307892.1 | 2 - 3 | 306 | 95 | ||
GenBank mRNA | AB169252.1 | - | 2 - 3 | 423 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |