Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283999.1 | NP_001270928.1 | 3 - 4 | 613 | 105 | |
XM_005559538.2 | XP_005559595.1 | 3 - 4 | 618 | 105 | ||
XM_005559539.2 | XP_005559596.1 | 3 - 4 | 617 | 105 | ||
XM_015452403.1 | XP_015307889.1 | 3 - 4 | 619 | 105 | ||
XM_015452404.1 | XP_015307890.1 | 3 - 4 | 613 | 105 | ||
XM_015452405.1 | XP_015307891.1 | 3 - 4 | 602 | 105 | ||
XM_015452406.1 | XP_015307892.1 | 3 - 4 | 496 | 105 | ||
XM_015452407.1 | XP_015307893.1 | 2 - 3 | 424 | 105 | ||
GenBank mRNA | AB169252.1 | - | 3 - 4 | 613 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |