Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACYP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285168.1 | NP_001272097.1 | 1 - 2 | 138 | 88 | |
XM_005575880.2 | XP_005575937.1 | 1 - 2 | 482 | 88 | ||
XM_015433439.1 | XP_015288925.1 | 1 - 2 | 482 | 88 | ||
XM_015433440.1 | XP_015288926.1 | 1 - 2 | 482 | 88 | ||
GenBank mRNA | AB172163.1 | - | 1 - 2 | 138 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |