Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005574979.2 | XP_005575036.1 | 2 - 3 | 622 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Inventoried Probe spans exons |