Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005540997.2 | XP_005541054.1 | 2 - 3 | 290 | 87 | |
GenBank mRNA | AB169197.1 | - | 2 - 3 | 234 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |