Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005590069.2 | XP_005590126.1 | 2 - 3 | 777 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |