Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015429889.1 | XP_015285375.1 | 2 - 3 | 1040 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |