Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005551339.2 | XP_005551396.1 | 2 - 3 | 187 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |