Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005595976.2 | XP_005596033.2 | 2 - 3 | 1198 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |