Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUGCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005549742.2 | XP_005549799.1 | 11 - 12 | 1026 | 79 | |
XM_005549745.2 | XP_005549802.1 | 11 - 12 | 1026 | 79 | ||
XM_015447507.1 | XP_015302993.1 | 11 - 12 | 1112 | 79 | ||
XM_015447509.1 | XP_015302995.1 | 11 - 12 | 1060 | 79 | ||
XM_015447510.1 | XP_015302996.1 | 11 - 12 | 1026 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |