Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102126088 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005568113.2 | XP_005568170.1 | 3 - 4 | 1269 | 125 | |
XM_005568115.2 | XP_005568172.1 | 3 - 4 | 1269 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons |