Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPH4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005551656.2 | XP_005551713.1 | 1 - 2 | 1178 | 102 | |
XM_005551657.2 | XP_005551714.1 | 1 - 2 | 1178 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |