Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005558731.2 | XP_005558788.1 | 3 - 4 | 278 | 61 | |
XM_005558733.2 | XP_005558790.1 | 3 - 4 | 562 | 61 | ||
XM_005558734.2 | XP_005558791.1 | 2 - 3 | 435 | 61 | ||
XM_015452355.1 | XP_015307841.1 | 2 - 3 | 541 | 61 | ||
XM_015452360.1 | XP_015307846.1 | 1 - 2 | 637 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Probe spans exons |