Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005588865.2 | XP_005588922.1 | 3 - 4 | 3713 | 85 | |
XM_015441696.1 | XP_015297182.1 | 4 - 5 | 2759 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |