Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEACAM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005589542.2 | XP_005589599.1 | 2 - 3 | 961 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |