Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMYD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005582455.2 | XP_005582512.1 | 4 - 5 | 497 | 65 | |
XM_005582456.2 | XP_005582513.1 | 4 - 5 | 640 | 65 | ||
XM_005582457.2 | XP_005582514.1 | 4 - 5 | 793 | 65 | ||
XM_005582458.2 | XP_005582515.1 | 4 - 5 | 399 | 65 | ||
XM_005582460.2 | XP_005582517.1 | 3 - 4 | 996 | 65 | ||
XM_005582461.2 | XP_005582518.1 | 2 - 3 | 194 | 65 | ||
XM_015437437.1 | XP_015292923.1 | 4 - 5 | 756 | 65 | ||
XM_015437438.1 | XP_015292924.1 | 4 - 5 | 756 | 65 | ||
XM_015437439.1 | XP_015292925.1 | 4 - 5 | 756 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |