Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287620.1 | NP_001274549.1 | 1 | 924 | 101 | |
XM_015445353.1 | XP_015300839.1 | 2 | 1422 | 101 | ||
GenBank mRNA | JX431563.1 | - | 1 | 924 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Inventoried Both primers and probe map within a single exon |