Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007537.1 | NP_031563.1 | 1 - 2 | 84 | 74 | |
GenBank mRNA | AF030769.1 | - | 1 - 2 | 84 | 74 | |
AK004680.1 | - | 1 - 2 | 114 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Detect Genomic DNA Mus musculus Probe spans exons |