Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ephb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146351.1 | NP_001139823.1 | 14 - 15 | 2943 | 73 | |
NM_007680.4 | NP_031706.3 | 14 - 15 | 2939 | 73 | ||
GenBank mRNA | AK076442.1 | - | 14 - 15 | 2766 | 73 | |
AK087423.1 | - | 14 - 15 | 2854 | 73 | ||
AK163823.1 | - | 5 - 6 | 661 | 73 | ||
BC031924.1 | - | 13 - 14 | 2551 | 4911959 | 73 | |
L77867.1 | - | 14 - 15 | 2633 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |