Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286033.1 | NP_001272962.1 | 6 - 7 | 613 | 67 | |
NM_001286034.1 | NP_001272963.1 | 6 - 7 | 608 | 67 | ||
NM_007941.3 | NP_031967.2 | 6 - 7 | 613 | 67 | ||
GenBank mRNA | AK146462.1 | - | 6 - 7 | 542 | 67 | |
AK165552.1 | - | 6 - 7 | 541 | 67 | ||
AK167577.1 | - | 6 - 7 | 576 | 67 | ||
AK168067.1 | - | 6 - 7 | 591 | 67 | ||
BC046279.1 | - | 6 - 7 | 537 | 6392250 | 67 | |
D10475.1 | - | 6 - 7 | 616 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |