Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grik5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008168.2 | NP_032194.2 | 18 - 19 | 2779 | 85 | |
GenBank mRNA | BC052009.1 | - | 18 - 19 | 2667 | 5695215 | 85 |
BC058097.1 | - | 18 - 19 | 2538 | 85 | ||
BC110682.1 | - | 18 - 19 | 2702 | 5033584 | 85 | |
D10011.1 | - | 18 - 19 | 2708 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |