Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008406.3 | NP_032432.2 | 5 - 6 | 584 | 76 | |
GenBank mRNA | AK149500.1 | - | 5 - 6 | 585 | 76 | |
BC013465.1 | - | 5 - 6 | 568 | 76 | ||
BC028814.1 | - | 5 - 6 | 584 | 5099284 | 76 | |
X70391.1 | - | 5 - 6 | 564 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |