Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Met | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008591.2 | NP_032617.2 | 13 - 14 | 3085 | 80 | |
GenBank mRNA | BC117969.1 | - | 11 - 12 | 2729 | 80 | |
BC117970.1 | - | 11 - 12 | 2725 | 80 | ||
CK332973.1 | - | 2 - 3 | 201 | 80 | ||
Y00671.1 | - | 11 - 12 | 2730 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |