Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mybpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008653.2 | NP_032679.2 | 23 - 24 | 2340 | 54 | |
GenBank mRNA | AF097333.1 | - | 23 - 24 | 2291 | 54 | |
AK146661.1 | - | 23 - 24 | 2341 | 54 | ||
AK146886.1 | - | 23 - 24 | 2341 | 54 | ||
AK169302.1 | - | 22 - 23 | 2318 | 54 | ||
AK169306.1 | - | 22 - 23 | 2318 | 54 | ||
BC094945.1 | - | 23 - 24 | 2337 | 54 | ||
BC131672.1 | - | 23 - 24 | 2316 | 54 | ||
BC154408.1 | - | 23 - 24 | 2361 | 40110890 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |