Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008939.2 | NP_032965.1 | 8 - 9 | 1821 | 92 | |
GenBank mRNA | BC031429.1 | - | 8 - 9 | 1723 | 3665834 | 92 |
D89871.1 | - | 8 - 9 | 1819 | 92 | ||
Y13192.1 | - | 8 - 9 | 1598 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |