Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008992.2 | NP_033018.2 | 9 - 10 | 1089 | 65 | |
GenBank mRNA | AJ001166.1 | - | 9 - 10 | 993 | 65 | |
AK028420.1 | - | 9 - 10 | 1127 | 65 | ||
AK183074.1 | - | 2 - 3 | 193 | 65 | ||
BC050102.1 | - | 9 - 10 | 1197 | 30012556 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |