Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009196.4 | NP_033222.1 | 3 - 4 | 571 | 97 | |
GenBank mRNA | AF058055.1 | - | 3 - 4 | 552 | 97 | |
AK033516.1 | - | 3 - 4 | 569 | 97 | ||
AK079379.1 | - | 3 - 4 | 567 | 97 | ||
AK088762.1 | - | 3 - 4 | 569 | 97 | ||
AK161701.1 | - | 3 - 4 | 569 | 97 | ||
AK162682.1 | - | 3 - 4 | 568 | 97 | ||
AK166606.1 | - | 3 - 4 | 569 | 97 | ||
BC014777.1 | - | 3 - 4 | 579 | 3499773 | 97 | |
X82438.1 | - | 2 - 3 | 363 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |