Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009209.3 | NP_033235.3 | 7 - 8 | 1349 | 98 | |
GenBank mRNA | AJ439987.1 | - | 7 - 8 | 1166 | 98 | |
AY188506.1 | - | 7 - 8 | 1166 | 98 | ||
AY525777.1 | - | 7 - 8 | 1463 | 98 | ||
AY525778.1 | - | 7 - 8 | 1463 | 98 | ||
BC066216.1 | - | 8 - 9 | 1283 | 30431573 | 98 | |
U76306.1 | - | 7 - 8 | 1148 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |