Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009294.3 | NP_033320.1 | 2 - 3 | 366 | 64 | |
GenBank mRNA | AK145909.1 | - | 2 - 3 | 385 | 64 | |
AK161971.1 | - | 2 - 3 | 359 | 64 | ||
BC005791.1 | - | 2 - 3 | 206 | 3603665 | 64 | |
BC011491.1 | - | 2 - 3 | 366 | 4016871 | 64 | |
U76832.1 | - | 2 - 3 | 135 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |