Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfpi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009364.3 | NP_033390.1 | 3 - 4 | 482 | 70 | |
GenBank mRNA | AK132268.1 | - | 3 - 4 | 504 | 70 | |
BC021639.1 | - | 3 - 4 | 517 | 4023742 | 70 | |
D50586.1 | - | 3 - 4 | 487 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |