Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Axl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190974.1 | NP_001177903.1 | 5 - 6 | 903 | 88 | |
NM_001190975.1 | NP_001177904.1 | 5 - 6 | 903 | 88 | ||
NM_009465.4 | NP_033491.2 | 5 - 6 | 903 | 88 | ||
GenBank mRNA | AK033237.1 | - | 5 - 6 | 872 | 88 | |
AK155567.1 | - | 6 - 7 | 789 | 88 | ||
AK170010.1 | - | 5 - 6 | 877 | 88 | ||
AK170897.1 | - | 6 - 7 | 800 | 88 | ||
BC046618.1 | - | 5 - 6 | 740 | 6494383 | 88 | |
BC050914.1 | - | 5 - 6 | 711 | 6418252 | 88 | |
BC058230.1 | - | 6 - 7 | 791 | 6313662 | 88 | |
X63535.1 | - | 5 - 6 | 652 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |