Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009521.2 | NP_033547.1 | 3 - 4 | 681 | 76 | |
GenBank mRNA | AK043014.1 | - | 2 - 3 | 510 | 76 | |
AK161781.1 | - | 1 - 2 | 78 | 76 | ||
AK163878.1 | - | 2 - 3 | 483 | 76 | ||
BC141271.1 | - | 3 - 4 | 606 | 9055883 | 76 | |
BC141274.1 | - | 3 - 4 | 606 | 9055886 | 76 | |
BC152304.1 | - | 3 - 4 | 617 | 40039305 | 76 | |
BC152305.1 | - | 3 - 4 | 618 | 40039312 | 76 | |
BC152537.1 | - | 3 - 4 | 618 | 40040396 | 76 | |
M32502.1 | - | 3 - 4 | 632 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |