Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009528.3 | NP_033554.3 | 1 - 2 | 442 | 88 | |
GenBank mRNA | BC052018.1 | - | 1 - 2 | 203 | 6401357 | 88 |
BC058398.1 | - | 1 - 2 | 442 | 88 | ||
M89802.1 | - | 1 - 2 | 71 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Mouse Wnt Panel Inventoried Probe spans exons |