Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bub1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009773.3 | NP_033903.2 | 7 - 8 | 1185 | 64 | |
GenBank mRNA | AF107296.1 | - | 7 - 8 | 1140 | 64 | |
AK133909.1 | - | 7 - 8 | 1126 | 64 | ||
BC031577.1 | - | 7 - 8 | 1136 | 4953223 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |