Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd3g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009850.2 | NP_033980.1 | 2 - 3 | 192 | 72 | |
GenBank mRNA | AK134049.1 | - | 2 - 3 | 125 | 72 | |
AK153980.1 | - | 2 - 3 | 124 | 72 | ||
AK153987.1 | - | 2 - 3 | 111 | 72 | ||
BC027528.1 | - | 2 - 3 | 123 | 1494674 | 72 | |
EF474453.1 | - | 2 - 3 | 81 | 72 | ||
GU456630.1 | - | 2 - 3 | 81 | 72 | ||
Y00635.1 | - | 2 - 3 | 220 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |