Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037801.2 | NP_001032890.1 | 5 - 6 | 1283 | 115 | |
NM_009852.3 | NP_033982.3 | 5 - 6 | 1283 | 115 | ||
GenBank mRNA | AK030822.1 | - | 5 - 6 | 1253 | 115 | |
AK030823.1 | - | 5 - 6 | 1284 | 115 | ||
AK088050.1 | - | 5 - 6 | 1261 | 115 | ||
BC014294.1 | - | 5 - 6 | 1289 | 4187689 | 115 | |
U12434.1 | - | 5 - 6 | 1081 | 115 | ||
U35370.1 | - | 5 - 6 | 1199 | 115 | ||
U37543.1 | - | 5 - 6 | 1240 | 115 | ||
U37544.1 | - | 5 - 6 | 1240 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |