Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145820.1 | NP_001139292.1 | 4 - 5 | 716 | 69 | |
NM_010274.3 | NP_034404.3 | 4 - 5 | 591 | 69 | ||
GenBank mRNA | AK034353.1 | - | 4 - 5 | 500 | 69 | |
AK144716.1 | - | 4 - 5 | 717 | 69 | ||
AK151189.1 | - | 4 - 5 | 503 | 69 | ||
AK162556.1 | - | 4 - 5 | 479 | 69 | ||
AK167152.1 | - | 4 - 5 | 529 | 69 | ||
BC021359.1 | - | 4 - 5 | 685 | 5012142 | 69 | |
D50430.1 | - | 4 - 5 | 493 | 69 | ||
U60987.1 | - | 4 - 5 | 527 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |