Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il6st | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010560.3 | NP_034690.3 | 10 - 11 | 1539 | 93 | |
GenBank mRNA | BC058679.1 | - | 10 - 11 | 1500 | 6834623 | 93 |
M83336.1 | - | 9 - 10 | 1344 | 93 | ||
X62646.1 | - | 9 - 10 | 1344 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |