Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010575.2 | NP_034705.2 | 26 - 27 | 2881 | 63 | |
GenBank mRNA | AF045019.1 | - | 3 - 4 | 345 | 63 | |
AF170316.1 | - | 26 - 27 | 2742 | 63 | ||
AK140749.1 | - | 21 - 22 | 2533 | 63 | ||
AK142289.1 | - | 26 - 27 | 2743 | 63 | ||
AK154619.1 | - | 26 - 27 | 2882 | 63 | ||
AK156872.1 | - | 25 - 26 | 3062 | 63 | ||
AK157012.1 | - | 25 - 26 | 3066 | 63 | ||
AK158443.1 | - | 26 - 27 | 2733 | 63 | ||
AK161727.1 | - | 5 - 6 | 757 | 63 | ||
BC120493.1 | - | 26 - 27 | 2731 | 40129416 | 63 | |
BC137570.1 | - | 26 - 27 | 2731 | 8860589 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |