Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190830.1 | NP_001177759.1 | 2 - 3 | 397 | 82 | |
NM_010589.6 | NP_034719.2 | 2 - 3 | 397 | 82 | ||
GenBank mRNA | AK038268.1 | - | 1 - 2 | 208 | 82 | |
AK088365.1 | - | 2 - 3 | 249 | 82 | ||
AK142178.1 | - | 2 - 3 | 235 | 82 | ||
AK156646.1 | - | 2 - 3 | 219 | 82 | ||
AK164520.1 | - | 2 - 3 | 237 | 82 | ||
BC027234.1 | - | 228 | 82 | |||
BC105577.1 | - | 1 - 2 | 200 | 3968386 | 82 | |
BC131646.1 | - | 1 - 2 | 208 | 40110146 | 82 | |
BC131647.1 | - | 1 - 2 | 208 | 40110148 | 82 | |
L40172.1 | - | 2 - 3 | 258 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |