Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011076.2 | NP_035206.2 | 16 - 17 | 2184 | 68 | |
GenBank mRNA | AY864315.1 | - | 15 - 16 | 2047 | 68 | |
M24417.1 | - | 10 - 11 | 1533 | 68 | ||
M30697.1 | - | 16 - 17 | 2198 | 68 | ||
M33581.1 | - | 16 - 17 | 2183 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mus musculus Probe spans exons |