Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291434.1 | NP_001278363.1 | 2 - 3 | 452 | 90 | |
NM_011102.4 | NP_035232.1 | 2 - 3 | 452 | 90 | ||
GenBank mRNA | AK144419.1 | - | 2 - 3 | 454 | 90 | |
BC111807.1 | - | 2 - 3 | 339 | 40039740 | 90 | |
L28035.1 | - | 2 - 3 | 530 | 90 | ||
X67129.1 | - | 2 - 3 | 261 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |